Learn More
Ace Glass, Inc. 20MM POR E TUBES (GB)
SDP
Catalog No. 50974378
Encompass_Preferred
20mm straight tubes with 4-8 micron fritted discs, for construction of special apparatus.
Catalog No. 50-974-378 Supplier Ace Glass, Inc. Supplier No. 720520
20mm straight tubes with 4-8 micron fritted discs, for construction of special apparatus.