Learn More
Chemglass Life Sciences 20mm Quartz Fritted Disc, Coarse
SDP
Catalog No. CGQ020708
Encompass_Preferred
20mm Quartz Fritted Disc, Coarse. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020708 Supplier Chemglass Life Sciences Supplier No. CGQ020708
20mm Quartz Fritted Disc, Coarse. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.