Learn More
Chemglass Life Sciences 20mm Quartz Fritted Disc, Medium
SDP
Catalog No. CGQ020707
Encompass_Preferred
20mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.
Catalog No. CGQ020707 Supplier Chemglass Life Sciences Supplier No. CGQ020707
20mm Quartz Fritted Disc, Medium. Porous fused quartz discs. Porosity sizes are as listed and differ from porosities of borosilicate discs.