Learn More
Analytical Sales & Services Advantage 20mm, 6mL Volume, 22 x 46mm, Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Round Bottom
Catalog No. 50454924
Encompass
Advantage 20mm, 6mL Volume, 22 x 46mm, Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Round Bottom
Catalog No. 50-454-924 Supplier Analytical Sales & Services Supplier No. 622823CASE
Advantage 20mm, 6mL Volume, 22 x 46mm, Clear Borosilicate Glass Crimp Head Space Vial with Beveled Top and Round Bottom