Learn More
West Pharmaceutical Services Inc 20mm Serum FluroTec™ Barrier Film-Laminated Ready-to-Sterilize Bromobutyl 4023/50 Stoppers with Envision™ Verification Process, Qty. 3,000 stoppers/bag
Catalog No. NC3931002
Encompass
FluroTec™ barrier film-laminated serum stoppers are made from 4023/50 a bromobutyl elastomer and laminated with FluroTec™ barrier film to reduce the risk of a chemical interaction. These components are provided washed (ready-to-sterilize) and 100% vision inspected. This stopper configuration can be used with European blowback, North American blowback, or straight wall (no blowback) vials. West item number:10145417.
Catalog No. NC3931002 Supplier West Pharmaceutical Services Inc Supplier No. 10145417
FluroTec™ barrier film-laminated serum stoppers are made from 4023/50 a bromobutyl elastomer and laminated with FluroTec™ barrier film to reduce the risk of a chemical interaction. These components are provided washed (ready-to-sterilize) and 100% vision inspected. This stopper configuration can be used with European blowback, North American blowback, or straight wall (no blowback) vials. West item number:10145417.