Learn More
Teknova 20mM Sodium Phosphate, 400mM Sodium Chloride, pH 7.0, 1000mL, Endotoxin Tested, Sterile. 8 pack.
SDP
Catalog No. 502745998
Encompass
20mM Sodium Phosphate, 400mM Sodium Chloride, pH 7.0, 1000mL, Endotoxin Tested, Sterile. 8 pack.
Catalog No. 50-274-5998 Supplier Teknova Supplier No. S24028PK
20mM Sodium Phosphate, 400mM Sodium Chloride, pH 7.0, 1000mL, Endotoxin Tested, Sterile. 8 pack.