Learn More
Agilent Technologies 20MM STEEL CRIMP CAP
Catalog No. NC1634442
Encompass
20mm steel crimp cap with a high temperature certified 20mm septum. 100/pack
Catalog No. NC1634442 Supplier Agilent Technologies Supplier No. 51903987
20mm steel crimp cap with a high temperature certified 20mm septum. 100/pack