Learn More
FENWAL INC 20mm ViaLok Vented 100/CS
Catalog No. 501961850
Encompass
• For use with Lovo version 3.0 software and product code X6R4909• Plastic spike, 0.2 μm vent, and female luer with cap• For needle-free vial access• Sterile fluid path• Individually packaged in form fill seal pouchVialok is a medical device and can be sold only in the US and the EU.
Catalog No. 50-196-1850 Supplier FENWAL INC Supplier No. YM020
• For use with Lovo version 3.0 software and product code X6R4909• Plastic spike, 0.2 μm vent, and female luer with cap• For needle-free vial access• Sterile fluid path• Individually packaged in form fill seal pouchVialok is a medical device and can be sold only in the US and the EU.