Learn More
Malvern Panalytical 20 Nm/15 mL Polystyrene Standard
Catalog No. NC2008024
Encompass
Nanosphere Size Standards 20nm /15ml.Nanosphere Size Standards are highly uniform polystyrene spheres calibrated by NIST traceable standards and packaged as aqueous suspensions in dropper-tipped bottles. 1% solids, optimized for easy dispersal and colloidal stability. Includes Certificate of Calibration and Traceability to NIST.
Catalog No. NC2008024 Supplier Malvern Panalytical Supplier No. 10033814
Nanosphere Size Standards 20nm /15ml.Nanosphere Size Standards are highly uniform polystyrene spheres calibrated by NIST traceable standards and packaged as aqueous suspensions in dropper-tipped bottles. 1% solids, optimized for easy dispersal and colloidal stability. Includes Certificate of Calibration and Traceability to NIST.