Learn More
Abcam 20nm Gold Nanoparticles (10 OD), 100ML
Catalog No. 502656254
Encompass_Preferred
20nm Gold Nanoparticles (10 OD) (ab269936) is an aqueous suspension of spherical metallic nanoparticles.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-6254 Supplier Abcam Supplier No. AB269936100ML
20nm Gold Nanoparticles (10 OD) (ab269936) is an aqueous suspension of spherical metallic nanoparticles.
The product is subject to the following: Abcam Restricted Use Statement