Learn More
Abcam 20nm Gold Nanoparticles (15 OD), 100ML
Catalog No. 502662813
Encompass_Preferred
20nm Gold Nanoparticles (15 OD) (ab269937) is an aqueous suspension of spherical metallic nanoparticles.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-2813 Supplier Abcam Supplier No. AB269937100ML
20nm Gold Nanoparticles (15 OD) (ab269937) is an aqueous suspension of spherical metallic nanoparticles.
The product is subject to the following: Abcam Restricted Use Statement