Learn More
Medchemexpress LLC (20R)-Ginsenoside Rh1 | 80952-71-2 | 638.87 | 20 MG
SDP
Catalog No. 5000414791
Encompass_Preferred
(20R)-Ginsenoside Rh1 is an R isomer of Ginsenoside Rh1, isolated from Panax Ginseng. It functions by inhibiting the thrombin-induced conversion of fibrinogen to fibrin, making it suitable for various research applications.
  • R isomer of Ginsenoside Rh1
  • Isolated from Panax Ginseng
  • Inhibits thrombin-induced conversion of fibrinogen to fibrin
  • For research use only
Catalog No. 50-004-14791 Supplier Medchemexpress LLC Supplier No. HYN140020MG
(20R)-Ginsenoside Rh1 is an R isomer of Ginsenoside Rh1, isolated from Panax Ginseng. It functions by inhibiting the thrombin-induced conversion of fibrinogen to fibrin, making it suitable for various research applications.
  • R isomer of Ginsenoside Rh1
  • Isolated from Panax Ginseng
  • Inhibits thrombin-induced conversion of fibrinogen to fibrin
  • For research use only