Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (20R)-Ginsenoside Rh1 | 80952-71-2 | 638.87 | 5 MG
SDP

Catalog No. 5000415476
Encompass_Preferred

(20R)-Ginsenoside Rh1 is the R isomer of Ginsenoside Rh1, isolated from Panax Ginseng. It functions by inhibiting the thrombin-induced conversion of fibrinogen to fibrin.

  • Purity: 99.06%
  • Appearance: solid
  • Color: white to off-white
  • Inhibits the thrombin-induced conversion of fibrinogen to fibrin

Catalog No. 50-004-15476 Supplier Medchemexpress LLC Supplier No. HYN14005MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(20R)-Ginsenoside Rh1 is the R isomer of Ginsenoside Rh1, isolated from Panax Ginseng. It functions by inhibiting the thrombin-induced conversion of fibrinogen to fibrin.

  • Purity: 99.06%
  • Appearance: solid
  • Color: white to off-white
  • Inhibits the thrombin-induced conversion of fibrinogen to fibrin

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.