Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (20R)-ginsenoside Rh1 | 80952-71-2 | 99.1% | 638.87 g/mol | C36H62O9 | 10 MG
SDP

Catalog No. 5000295781
Encompass_Preferred

(20R)-Ginsenoside Rh1 is the R-isomer of ginsenoside Rh1 isolated from Panax ginseng and supplied as a purified solid for biochemical and analytical research. It is reported to inhibit the thrombin-induced conversion of fibrinogen to fibrin.

  • High purity (99.06%).
  • Molecular formula C36H62O9; molecular weight 638.87 g/mol.
  • Supplied as a solid suitable for use as a natural product standard.
  • Storage: -20°C, protect from light; in solvent: -80°C for 6 months or -20°C for 1 month (protect from light).
  • Common applications include natural product research, biochemical assays, and use as an analytical reference standard.
  • Available in small laboratory pack sizes (5 mg-100 mg).

Catalog No. 50-002-95781 Supplier Medchemexpress LLC Supplier No. HYN140010MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(20R)-Ginsenoside Rh1 is the R-isomer of ginsenoside Rh1 isolated from Panax ginseng and supplied as a purified solid for biochemical and analytical research. It is reported to inhibit the thrombin-induced conversion of fibrinogen to fibrin.

  • High purity (99.06%).
  • Molecular formula C36H62O9; molecular weight 638.87 g/mol.
  • Supplied as a solid suitable for use as a natural product standard.
  • Storage: -20°C, protect from light; in solvent: -80°C for 6 months or -20°C for 1 month (protect from light).
  • Common applications include natural product research, biochemical assays, and use as an analytical reference standard.
  • Available in small laboratory pack sizes (5 mg-100 mg).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.