Learn More
Medchemexpress LLC HY-N2040 5mg Medchemexpress, (20R)-Protopanaxadiol CAS:7755-01-3 Purity:>98%
SDP
Catalog No. 502082860
Encompass_Preferred
Medchemexpress, HY-N2040 5mg (20R)-Protopanaxadiol CAS:7755-01-3 (20R)-Protopanaxadiol is a triterpenoid saponin metabolite of 20(R)-ginsenoside Rg3 in black ginseng. (20R)-Protopanaxadiol exhibits anti-tumor activity and cytotoxicity, and potently inhibits the growth of Helicobacter pylori. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2860 Supplier Medchemexpress LLC Supplier No. HYN20405MG
Medchemexpress, HY-N2040 5mg (20R)-Protopanaxadiol CAS:7755-01-3 (20R)-Protopanaxadiol is a triterpenoid saponin metabolite of 20(R)-ginsenoside Rg3 in black ginseng. (20R)-Protopanaxadiol exhibits anti-tumor activity and cytotoxicity, and potently inhibits the growth of Helicobacter pylori. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.