Learn More
Lkt Laboratories 20S-Ginsenoside Rg3, ≥98%, 5MG
SDP
Supplier:  Lkt Laboratories G35525MG
Encompass_Preferred
Triterpene saponin found in Panax; Kv7.1 K+ channel activator, γ2 GABA-A agonist, α10 nAChR antagonist. CAS: 14197-60-5. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4925