Learn More
Enzo Life Sciences Proteasome 20S alpha1, 2, 3, 5, 6 & 7 subunits monoclonal antibody (MCP231) (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLPW81950025
Encompass_Preferred
Clone: MCP231. Host: Mouse. Isotype: IgG1κ. Immunogen: Dinitrophenylated proteasomes. UniProt: P25786 (human PSMA1), P25787 (human PSMA2), P25788 (human PSMA3), P25789 (human PSMA4), P28066 (human PSMA5), P60900 (human PSMA6). Source: Purified from ascites. Species Reactivity: Human, Mouse, Rat, Yeast, Rabbit, Potato. Specificity: Recognizes the α1, 2, 3, 5, 6 & 7 subunits of the 20S proteasome. Applications: IHC, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 0.01% sodium azide. Handling: Store unopened vial at -20°C until required for use. Aliquot after opening and store at 2-4°C. The use of high quaity \'antiserum-grade\' plastic or glass vials is recommended. Use within 1 month. Avoid freeze/thaw cycles.
Catalog No. NC0962410