Learn More
AdipoGen 20S Proteasome (mouse) (untagged) (50 µg)
SDP
Supplier:  AdipoGen SBBPP0047C050
Encompass_Preferred
Protein. Mouse 20S Proteasome. Source: Mouse Erythrocytes. Liquid. In 50mM HEPES pH 7.5, 100mM sodium chloride, 1mM TCEP. Purity: >95% (SDS-PAGE). The ubiquitin-proteasome pathway is the major proteolytic system in eukaryotic cells, where it catalyzes the selective degradation of short-lived regulatory proteins or the rapid turnover of misfolded proteins. One of the most important proteases in this pathway is the 26S proteasome, an ATP-dependent proteolytic complex, which is formed by the association of the barrel-shaped 20S proteasome (700kDa) and two 19S (700kDa) regulatory complexes. The 20S catalytic core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha-subunits and 2 rings are composed of 7 beta-subunits. The 20S catalytic core is able to degrade a variety of peptide substrates and poly-ubiquitinated proteins involved with apoptosis, DNA repair, endocytosis and cell cycle control.
Catalog No. 50-205-4700