Learn More
TARGETMOL CHEMICALS INC 20S-PROTOPANAXADIOL 50MG
SDP
Catalog No. 502580554
Encompass_Preferred
Also available in 5 mg 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (20S)-Protopanaxadiol (20-Epiprotopanaxadiol) (20-Epiprotopanaxadiol) an apoptosis inducer is an aglycon metabolic derivative of the protopanaxadiol-type ginseng saponin. purity: 98%
Catalog No. 50-258-0554 Supplier TARGETMOL CHEMICALS INC Supplier No. T279950MG
Also available in 5 mg 10 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (20S)-Protopanaxadiol (20-Epiprotopanaxadiol) (20-Epiprotopanaxadiol) an apoptosis inducer is an aglycon metabolic derivative of the protopanaxadiol-type ginseng saponin. purity: 98%