Learn More
Alkali Scientific Alkali Scientific™ 20uL Beckman™ Biomek FX/NX, Multimek and i Series Compatible Robotic Automation Black Conductive Tips (P20), 96-Well Head/Tip Format, Low Retention, Sterile, Filtered, 10 Racks of 96, 960/Pack
SDP
Supplier:  Alkali Scientific FXBF20
Encompass
  Crafted from high-quality polypropylene, Alkali Scientific™ offers an extensive selection of tips for Beckman Coulter automation systems. These black conductive tips are produced under strict specifications and process controls, with automated assembly to ensure reliable performance and consistency. The tips are certified to be free of RNase, DNase, and pyrogens. They come in various configurations, including nonsterile, sterile, non-filtered, filtered, wide-bore, fine point, and extended length options. Compatible with Beckman Coulter Biomek FX, Biomek NX, Biomek 3000, Multimek AP-96, i-Series, SAP96 Head, Span-8, P20, MP20, P200L, and MP200. Filtered tips help minimize the risk of sample cross-contamination due to aerosols. All P20 tips are made from 100% polypropylene, ensuring they are straight, leak-proof, and guaranteed to be free from DNA, RNase/DNase, PCR inhibitors, pyrogens/endotoxins, and trace metals.
Catalog No. 50-302-8727