Learn More
Genesee Scientific Corporation 20µL Classic Filter Pipette Tips, Low Binding, Ergonomic Design, Sterile
SDP
Catalog No. NC1959564
Encompass
The Genesee Scientific 20µL Classic Filter Pipette Tips are designed for precise and efficient liquid handling. Featuring an ergonomic design and naturally low-retentive resin, they reduce sample retention and enhance recovery. Key benefits include color-coded inserts for easy identification, certified free from contamination, and a design that minimizes hanging droplets, promoting reliability and ease of use in laboratory workflows.
Catalog No. NC1959564 Supplier Genesee Scientific Corporation Supplier No. 24404C
The Genesee Scientific 20µL Classic Filter Pipette Tips are designed for precise and efficient liquid handling. Featuring an ergonomic design and naturally low-retentive resin, they reduce sample retention and enhance recovery. Key benefits include color-coded inserts for easy identification, certified free from contamination, and a design that minimizes hanging droplets, promoting reliability and ease of use in laboratory workflows.