Learn More
Trajan Scientific VICI Valco Sample Loop 20-L Cheminert (VAL.CSL20)
Catalog No. 50635008
Encompass
VICI Valco Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and ferrµles. VICI Valco Part Number: CSL20 (NEW LPP PN: VAL.CSL20). Thermo Part Number: CH-953543. Made in the USA.
Catalog No. 50-635-008 Supplier Trajan Scientific Supplier No. PAL.CSL20
VICI Valco Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and ferrµles. VICI Valco Part Number: CSL20 (NEW LPP PN: VAL.CSL20). Thermo Part Number: CH-953543. Made in the USA.