Learn More
Genesee Scientific Corporation Olympus 20 µL Barrier Tips, RNase Free, Sterile, 20 µL
SDP
Catalog No. NC1962412
Encompass
The Olympus 20 µL Barrier Tips provide precision-manufactured pipette tips that ensure accurate and contaminant-free liquid handling in scientific applications. Constructed from virgin polypropylene with a unique low-binding surface, these sterile tips minimize sample retention to enhance retrieval accuracy. They are RNase, DNase, DNA, and pyrogen-free, making them ideal for sensitive assays. The universal fit design allows compatibility with most research-grade pipettors, promoting laboratory efficiency.
Catalog No. NC1962412 Supplier Genesee Scientific Corporation Supplier No. 26404
The Olympus 20 µL Barrier Tips provide precision-manufactured pipette tips that ensure accurate and contaminant-free liquid handling in scientific applications. Constructed from virgin polypropylene with a unique low-binding surface, these sterile tips minimize sample retention to enhance retrieval accuracy. They are RNase, DNase, DNA, and pyrogen-free, making them ideal for sensitive assays. The universal fit design allows compatibility with most research-grade pipettors, promoting laboratory efficiency.