Learn More
Alkali Scientific 20uL PerkinElmer Revvity Janus
SDP
Supplier:  Alkali Scientific PEST1020
Encompass_Preferred
Alkali Scientifichas designed its PerkinElmer automation tips for optimal compatibility with PerkinElmer Janus Varispan liquid handlers With our PerkinElmer Janus-compatible tips no separate labware definition is required and theres no need to modify PerkinElmer Janus automation control software protocols These 96-format conductive black tips are fully interchangeable with the original PerkinElmer P20 automation pipette tips and are also compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems ApplicationsDesigned for volume transfers up to 20 L the PerkinElmer P20 conductive black tip delivers exceptional precision and accuracy Its volume capacity and length provide flexibility for protocol development These tips can access both 96-format and 384-format plates and are compatible with Janus Varispan pipetting arms Alkali Scientificmanufactures its PerkinElmer Janus-compatible P20 tips using premium virgin polypropylene We employ advanced
Catalog No. 50-302-9283