Learn More
Alkali Scientific 20uL PerkinElmer Revvity Janus
SDP
Supplier:  Alkali Scientific PEBST1020
Encompass_Preferred
Our designs its PerkinElmer automation tips for optimal compatibility with PerkinElmer Janus Varispan liquid handlers When using our PerkinElmer Janus-compatible automation tips theres no need for a separate labware definition and the PerkinElmer Janus automation control software protocols remain unchanged These 96-format conductive black tips are fully interchangeable with the original PerkinElmer P20 automation pipette tips They are also compatible with Sciclone Zephyr Evolution P3 RapidPlate MiniTrak and MultiProbe II HT/ET systems ApplicationsThese 96-format tips designed for volume transfers up to 20 L offer exceptional precision and accuracy The volume capacity and length provide flexibility in protocol design and they can be used to access both 96-format and 384-format plates These tips are compatible with the Janus Varispan pipetting arms Alkali Scientificmanufactures its PerkinElmer Janus-compatible P20 tips from high-quality virgin polypropylene We employ ultra-poli
Catalog No. 50-302-8307