Learn More
Teknova 20X HBS-EP+ Buffer, pH 7.6.1000mL, Sterile.
SDP
Catalog No. NC0124284
Encompass
3M Sodium Chloride, 200mM HEPES Free Acid,60mM EDTA, 1.0% Tween-20
This item is not returnable. View return policy
Catalog No. NC0124284 Supplier Teknova Supplier No. H8022
3M Sodium Chloride, 200mM HEPES Free Acid,60mM EDTA, 1.0% Tween-20