Learn More
Teknova 20X PBS. 20L, Sterile
SDP
Catalog No. 502387664
Encompass
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.
Catalog No. 50-238-7664 Supplier Teknova Supplier No. P1225
Phosphate-buffered saline (PBS) is a salt solution that is commonly used in physiological and biochemical experiments. It can be used to maintain pH neutrality or to buffer against changes in pH in order to provide optimal conditions for cellular growth and function. The buffering range is between pH 6.8 and 7.4.