Learn More
Boston Bioproducts Inc Phosphate Buffered Saline, PBS (20X) - 500ml
Catalog No. NC1430371
Encompass
Phosphate Buffered Saline (PBS) is a buffer solution widely used in biological research due to its isotonic and non-toxic nature. With a pH range of 7.0 to 7.4, PBS mimics the osmolarity and ion concentrations of the human body, making it ideal for applications including protein purification, Western Blotting and immunoassays, sample dilution, and as a transport medium. This buffer is formulated in a 20X concentration, and can be diluted to support its application.
Catalog No. NC1430371 Supplier Boston Bioproducts Inc Supplier No. BM220X500ML
Phosphate Buffered Saline (PBS) is a buffer solution widely used in biological research due to its isotonic and non-toxic nature. With a pH range of 7.0 to 7.4, PBS mimics the osmolarity and ion concentrations of the human body, making it ideal for applications including protein purification, Western Blotting and immunoassays, sample dilution, and as a transport medium. This buffer is formulated in a 20X concentration, and can be diluted to support its application.