Learn More
Boston Bioproducts Inc Phosphate Buffered Saline-Tween, PBST (20X, with 1% Tween-20, pH 7.4) - 1L
Catalog No. NC9396008
Encompass
20X Phosphate Buffered Saline with Tween-20 (PBST) is a specialized variant of PBS, supplemented with Tween-20, a non-ionic surfactant. Tween-20 minimizes the non-specific binding and background noise, resuting in enhanced signal to noise ratio. PBST is often used in immunoassays such as ELISA and Western Blotting, where it serves as a washing and blocking buffer. With a stable buffering pH range between 7.0 and 7.4, PBST maintains ionic strength and protein solubility for biological assays.
Catalog No. NC9396008 Supplier Boston Bioproducts Inc Supplier No. IBB171X1L
20X Phosphate Buffered Saline with Tween-20 (PBST) is a specialized variant of PBS, supplemented with Tween-20, a non-ionic surfactant. Tween-20 minimizes the non-specific binding and background noise, resuting in enhanced signal to noise ratio. PBST is often used in immunoassays such as ELISA and Western Blotting, where it serves as a washing and blocking buffer. With a stable buffering pH range between 7.0 and 7.4, PBST maintains ionic strength and protein solubility for biological assays.