Learn More
Seracare Life Sciences Inc 20X SSC
Catalog No. 5067447
Encompass_Preferred
20X SSC is a common buffer used in a variety of molecular biology applications. It is a clear, colorless solution made up of molecular biology grade 3.0 M NaCl and 0.3 M sodium citrate at pH 7.0. The solution is filtered through a 0.2 filter into a DNase/RNase free bottles. 20X SSC can be used to facilitate transfer of nucleic acids to membranes in Southern and Northern blotting, dot blotting, and colony and plaque lifts.
This item is not returnable. View return policy
Catalog No. 50-674-47 Supplier Seracare Life Sciences Inc Supplier No. 59600021
20X SSC is a common buffer used in a variety of molecular biology applications. It is a clear, colorless solution made up of molecular biology grade 3.0 M NaCl and 0.3 M sodium citrate at pH 7.0. The solution is filtered through a 0.2 filter into a DNase/RNase free bottles. 20X SSC can be used to facilitate transfer of nucleic acids to membranes in Southern and Northern blotting, dot blotting, and colony and plaque lifts.