Learn More
Growcells 20X SSC 2L EA
Catalog No. 501034860
Encompass_Preferred
Sterile 20X Standard Sodium Citrate (SSC) prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging.
Catalog No. 50-103-4860 Supplier Growcells Supplier No. MRGF6496
Sterile 20X Standard Sodium Citrate (SSC) prepared with 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging.