Learn More
Teknova 20X SSC Molecular Biology Grade, 10L, Non-Sterile.
SDP
Catalog No. 502387845
Encompass
3.0M Sodium Chloride,0.3M Citric Acid, Trisodium Salt, Adjusted to pH 7.0
Catalog No. 50-238-7845 Supplier Teknova Supplier No. S0283
3.0M Sodium Chloride,0.3M Citric Acid, Trisodium Salt, Adjusted to pH 7.0