Learn More
Rockland Immunochemicals 20X SSC pH 7.0 (3.0 M Sodium Chloride, 0.3 M Sodium Citrate, pH 7.0) DEPC TREATED - MB-045
SDP
Catalog No. 501059004
Encompass_Preferred
Saline-sodium citrate (SSC) buffer is most commonly used as a buffer for the hybridization process and washing in experiments where spcies of nucleic or ribonucleic acid are to be matched to complimentary sequences (such as Southern and Nouthern blotting, in situ hybridization, or DNA microarrays). SSC is used to help control the stringency of hybridization in these steps.
Catalog No. 50-105-9004 Supplier Rockland Immunochemicals Supplier No. MB045
Saline-sodium citrate (SSC) buffer is most commonly used as a buffer for the hybridization process and washing in experiments where spcies of nucleic or ribonucleic acid are to be matched to complimentary sequences (such as Southern and Nouthern blotting, in situ hybridization, or DNA microarrays). SSC is used to help control the stringency of hybridization in these steps.