Learn More
Teknova 20X Wash Buffer A, 1000mL, Sterile.
SDP
Catalog No. 501908097
Encompass
2.7M Sodium Chloride, 54mM Potassium Chloride, 86mM Disodium Phosphate, 28mM Monopotassium Phosphate,2% Tween-20
Catalog No. 50-190-8097 Supplier Teknova Supplier No. P2192
2.7M Sodium Chloride, 54mM Potassium Chloride, 86mM Disodium Phosphate, 28mM Monopotassium Phosphate,2% Tween-20