Learn More
EXTRACT TECHNOLOGY 20X20 AMMONIA LEAK DET CLOTH
SDP
Catalog No. NC0467571
Encompass
The Ammonia Leak Detection Cloth is used for the detection of leaks in sealed incubators, such as those used for infectious disease studies and in the treatment of premature birth. Industrial uses for detection of ammonia gas include sealed containers and filtration systems. The cloth is sensitive to vapors from a 1% solution of Ammonia.
This item is not returnable. View return policy
Catalog No. NC0467571 Supplier EXTRACT TECHNOLOGY Supplier No. 7030002303
The Ammonia Leak Detection Cloth is used for the detection of leaks in sealed incubators, such as those used for infectious disease studies and in the treatment of premature birth. Industrial uses for detection of ammonia gas include sealed containers and filtration systems. The cloth is sensitive to vapors from a 1% solution of Ammonia.