Learn More
Miles Scientific 20X20 GF UNIPLATE-T TAPER 25/B
SDP
Catalog No. NC9176859
Encompass_Preferred
Plate, TLC, Tapered, Preparative, Silica Gel GF, Gypsum Binder, Preadsorbent Zone, Uniplate-T, 20L x 20cmW
Catalog No. NC9176859 Supplier Miles Scientific Supplier No. P81013
Plate, TLC, Tapered, Preparative, Silica Gel GF, Gypsum Binder, Preadsorbent Zone, Uniplate-T, 20L x 20cmW