Learn More
Miles Scientific 20X20 GHLF 250U 25/BX
SDP
Catalog No. NC9940132
Encompass_Preferred
Silica Gel GHLF 250micrometer 20x20cm 25 plates/box
Catalog No. NC9940132 Supplier Miles Scientific Supplier No. P21011
Silica Gel GHLF 250micrometer 20x20cm 25 plates/box