Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 20mm ID X 22mm OD
SDP
Catalog No. CGQ031018
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 20mm ID X 22mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031018 Supplier Chemglass Life Sciences Supplier No. CGQ031018
Graded Seal, Quartz to Borosilicate, 20mm ID X 22mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.