This pleated air filter features a wire-backed design, synthetic media, and a MERV 10 rating for efficient particle capture. It is suitable for various HVAC applications and designed to meet industry standards.
- Wire-backed pleated design for enhanced structural integrity
- MERV 10 rating provides effective filtration against airborne particles
- Synthetic media material offers high efficiency and durability
- Paperboard frame ensures secure fit and easy installation
- Nominal size of 20 x 25 x 2 in. for broad compatibility
- Achieves 55% particle capture efficiency
- Rated for 1750 CFM airflow and a maximum velocity of 500 FPM
- Integrated frame allows for universal use with any unit
- Complies with ASHRAE 52.2-2012 and UL 900 standards
- Operates effectively up to 200 °F (93.3 °C)