Learn More
Kd Medical Inc 20XSSC(3M NACL0.3M SODCITR)4L
SDP
Catalog No. 501031439
Encompass_Preferred
Used in nucleic acid hybridization. 3M NaCl + 0.3M Sodium Citrate. One 4 liter cubitainer.
Catalog No. 50-103-1439 Supplier Kd Medical Inc Supplier No. RGH0240
Used in nucleic acid hybridization. 3M NaCl + 0.3M Sodium Citrate. One 4 liter cubitainer.