Learn More
Med Vet International 21 1/2" Elbow Length Poly Glove 100/Box
Catalog No. 502830361
Encompass_Preferred
Elbow length poly gloves are perfect for light tasks such as mixing salads or food in vats and bins in schools, bakeries, and restaurants. Each glove is made of clear .03 mm thick polyethylene and measures 21 1/2" long to provide plenty of protection for a worker's entire forearm and prevent food contamination. This poly glove is made of 100% latex free material to accommodate people with allergies to latex
Catalog No. 50-283-0361 Supplier Med Vet International Supplier No. AHSELGELBOW
Elbow length poly gloves are perfect for light tasks such as mixing salads or food in vats and bins in schools, bakeries, and restaurants. Each glove is made of clear .03 mm thick polyethylene and measures 21 1/2" long to provide plenty of protection for a worker's entire forearm and prevent food contamination. This poly glove is made of 100% latex free material to accommodate people with allergies to latex