Learn More
Medchemexpress LLC HY-136340 10mg Medchemexpress, 21-Acetoxypregna-1,4,9(11),16-tetraene-3,20-dione CAS:37413-91-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13634010MG
Encompass_Preferred
Medchemexpress, HY-136340 10mg 21-Acetoxypregna-1,4,9(11),16-tetraene-3,20-dione CAS:37413-91-5 21-Acetoxypregna-1,4,9(11),16-tetraene-3,20-dione is an intermediate of delta 9,11 steroids synthesis, for example, Vamorolone (HY-109017). The delta 9,11 steroids are modifications of glucocorticoids and has anti-inflammatory properties. The delta 9,11 steroids are agents for protection against cell damage (lipid peroxidation) and inhibition of neovascularization. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1437