Learn More
Lascar Electronics Inc High Accuracy Thermistor Probe Data Logger with LCD
SDP
Catalog No. NC1856625
Encompass_Preferred
This standalone data logger records and displays over 32,000 temperature readings from the supplied thermistor probe.
Catalog No. NC1856625 Supplier Lascar Electronics Inc Supplier No. ELUSBTPLCD+
This standalone data logger records and displays over 32,000 temperature readings from the supplied thermistor probe.