Learn More
Lascar Electronics Inc 21CFR USB High Accuracy Thermistor Probe Logger with Display
SDP
Catalog No. NC1856624
Encompass_Preferred
This 21CFR-compliant standalone data logger records and displays over 32,000 temperature readings from the supplied high accuracy thermistor probe. The data logger features an LCD and push-button which allow the user to cycle through the most recent, highest and lowest stored readings.
Catalog No. NC1856624 Supplier Lascar Electronics Inc Supplier No. EL21CFRTPLCD+
This 21CFR-compliant standalone data logger records and displays over 32,000 temperature readings from the supplied high accuracy thermistor probe. The data logger features an LCD and push-button which allow the user to cycle through the most recent, highest and lowest stored readings.