Learn More
Lascar Electronics Inc 21CFR WiFi Dual Channel Thermocouple Probe Data Logging Sensor
SDP
Catalog No. NC1856670
Encompass_Preferred
This 21CFR WiFi-enabled dual channel data logger can remotely monitor an environment using two inter-changeable J, K or T type thermocouples.
Catalog No. NC1856670 Supplier Lascar Electronics Inc Supplier No. ELWIFI21CFRDTC
This 21CFR WiFi-enabled dual channel data logger can remotely monitor an environment using two inter-changeable J, K or T type thermocouples.