Learn More
Lascar Electronics Inc 21CFR WiFi Thermocouple Data Logger
SDP
Catalog No. NC1856667
Encompass_Preferred
This 21CFR WiFi-enabled data logger can remotely monitor an environment using an inter-changeable J, K or T type thermocouple.
Catalog No. NC1856667 Supplier Lascar Electronics Inc Supplier No. ELWIFI21CFRTC
This 21CFR WiFi-enabled data logger can remotely monitor an environment using an inter-changeable J, K or T type thermocouple.