Learn More
Medchemexpress LLC 21-deoxy cortisone | 1882-82-2 | MFCD00051123 | 344.44 | C21H28O4 | 5mg
SDP
Catalog No. 5000270502
Encompass_Preferred
21-Deoxy Cortisone (21-Desoxycortisone NSC 38722) is a corticosteroid metabolite of 11-ketoprogesterone It is formed from 11-ketoprogesterone by the cytochrome P450 (CYP) isozyme CYP17A1 but can also be produced by oxidation of 21-deoxycortisone (HY-113405) by 11 -hydroxysteroid dehydrogenase type 2 (11 -HSD2) Congenital adrenal hyperplasia is an inborn error of metabolism characterized by a deficiency of 21-hydroxylase and 21-Deoxy Cortisone levels are elevated in patients with congenital adrenal hyperplasia
Catalog No. 50-002-70502 Supplier Medchemexpress LLC Supplier No. HYW7415105MG
21-Deoxy Cortisone (21-Desoxycortisone NSC 38722) is a corticosteroid metabolite of 11-ketoprogesterone It is formed from 11-ketoprogesterone by the cytochrome P450 (CYP) isozyme CYP17A1 but can also be produced by oxidation of 21-deoxycortisone (HY-113405) by 11 -hydroxysteroid dehydrogenase type 2 (11 -HSD2) Congenital adrenal hyperplasia is an inborn error of metabolism characterized by a deficiency of 21-hydroxylase and 21-Deoxy Cortisone levels are elevated in patients with congenital adrenal hyperplasia