Learn More
Cayman Chemical 21-Deoxycortisol, Purity: ≥95%, CAS: 641-77-0, 10 mg
SDP
Catalog No. 502988024
Encompass_Preferred
A corticosteroid metabolite of 17-hydroxyprogesterone; serum levels are elevated in patients with congenital adrenal hyperplasia that are heterozygous for mutations in CYP2A21 and have been used as a biomarker for the detection of 21-hydroxylase deficiencies
Catalog No. 50-298-8024 Supplier Cayman Chemical Supplier No. 3087310 MG
A corticosteroid metabolite of 17-hydroxyprogesterone; serum levels are elevated in patients with congenital adrenal hyperplasia that are heterozygous for mutations in CYP2A21 and have been used as a biomarker for the detection of 21-hydroxylase deficiencies