Learn More
Medchemexpress LLC 21-Deoxycortisol-d8 | 2479914-04-8 | 98.79% | 354.51 | 1 MG
SDP
Catalog No. 5000435805
Encompass_Preferred
21-Deoxycortisol-d8 is the deuterium labeled 21-Deoxycortisol. 21-Deoxycortisol is an endogenous metabolite and a sign of congenital adrenal hyperplasia. This compound can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-35805 Supplier Medchemexpress LLC Supplier No. HY113405S11MG
21-Deoxycortisol-d8 is the deuterium labeled 21-Deoxycortisol. 21-Deoxycortisol is an endogenous metabolite and a sign of congenital adrenal hyperplasia. This compound can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.