Learn More
Medchemexpress LLC 21-deoxycortisone | 1882-82-2 | MFCD00051123 | >95.0% | 344.44 | C21H28O4 | 10mg
SDP
Catalog No. 5000269615
Encompass_Preferred
21-Deoxy Cortisone (21-Desoxycortisone NSC 38722) is a corticosteroid metabolite of 11-ketoprogesterone It is formed from 11-ketoprogesterone by the cytochrome P450 (CYP) isozyme CYP17A1 but can also be produced by oxidation of 21-deoxycortisone (HY-113405) by 11 -hydroxysteroid dehydrogenase type 2 (11 -HSD2) Congenital adrenal hyperplasia is an inborn error of metabolism characterized by a deficiency of 21-hydroxylase and 21-Deoxy Cortisone levels are elevated in patients with congenital adrenal hyperplasia
Catalog No. 50-002-69615 Supplier Medchemexpress LLC Supplier No. HYW74151010MG
Add to Cart
Add to Cart
21-Deoxy Cortisone (21-Desoxycortisone NSC 38722) is a corticosteroid metabolite of 11-ketoprogesterone It is formed from 11-ketoprogesterone by the cytochrome P450 (CYP) isozyme CYP17A1 but can also be produced by oxidation of 21-deoxycortisone (HY-113405) by 11 -hydroxysteroid dehydrogenase type 2 (11 -HSD2) Congenital adrenal hyperplasia is an inborn error of metabolism characterized by a deficiency of 21-hydroxylase and 21-Deoxy Cortisone levels are elevated in patients with congenital adrenal hyperplasia